Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004427.3 | NP_004418.2 | 6 | 924 | |
NM_198040.2 | NP_932157.1 | 13 | 2231 | ||
XM_005270570.1 | XP_005270627.1 | 6 | 860 | ||
XM_017000513.1 | XP_016856002.1 | 14 | 2730 | ||
XM_017000514.1 | XP_016856003.1 | 14 | 2342 | ||
XM_017000515.1 | XP_016856004.1 | 14 | 2435 | ||
XM_017000516.1 | XP_016856005.1 | 14 | 2664 | ||
XM_017000517.1 | XP_016856006.1 | 14 | 2276 | ||
XM_017000518.1 | XP_016856007.1 | 14 | 2369 | ||
XM_017000519.1 | XP_016856008.1 | 14 | 2739 | ||
XM_017000520.1 | XP_016856009.1 | 13 | 2189 | ||
XM_017000521.1 | XP_016856010.1 | 13 | 2608 | ||
GenBank mRNA | AJ419231.1 | - | 13 | 2231 | |
AK056429.1 | - | 7 | 1161 | ||
AK128821.1 | - | 6 | 742 | ||
AK292905.1 | - | 14 | 2669 | ||
AK294426.1 | - | 8 | 1509 | ||
BC015450.1 | - | 2 | 71 | ||
BC018602.1 | - | 6 | 930 | ||
BC028396.1 | - | 6 | 895 | ||
BC029269.1 | - | 6 | 766 | ||
BC068573.1 | - | 6 | 818 | ||
BC092492.1 | - | 6 | 818 | ||
BC110863.1 | - | 6 | 817 | ||
BC130630.1 | - | 12 | 2147 | 40146661 | |
BC144121.1 | - | 12 | 2147 | 9052641 |