Gene Transcripts

Gene Symbol
FBLN1
Entrez Gene ID
2192
Gene Name
fibulin 1
Gene Aliases
FBLN, FIBL1
Chromosome Location
Chr. 22: 45502839 - 45601134 on Build GRCh38
UniGene ID
Hs.24601
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001996.3 NP_001987.2 8 1054
NM_006485.3 NP_006476.2 8 1054
NM_006486.2 NP_006477.2 8 1054
NM_006487.2 NP_006478.2 8 1054
GenBank mRNA AF126110.1 - 8 917
AF217999.1 - 8 997
AK075460.1 - 9 1124
AK075566.1 - 8 906
AK300806.1 - 8 990
AK316291.1 - 8 990
BC022497.1 - 8 1010 4819750
U01244.1 - 8 917
X53741.1 - 8 917
X53742.1 - 8 917
X53743.1 - 8 917

View Full Product Details