Gene Transcripts

Gene Symbol
FLNA
Entrez Gene ID
2316
Gene Name
filamin A
Gene Aliases
ABP-280, ABPX, CSBS, CVD1, FLN, FLN-A, FLN1, FMD, MNS, NHBP, OPD, OPD1, OPD2, XLVD, XMVD
Chromosome Location
Chr. X: 154348529 - 154374638 on Build GRCh38
UniGene ID
Hs.195464
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110556.1 NP_001104026.1 37 6125
NM_001456.3 NP_001447.2 36 6101
XM_011531127.1 XP_011529429.1 36 6024
XM_011531128.1 XP_011529430.1 35 6000
XM_011531129.1 XP_011529431.1 36 5946
XM_011531130.1 XP_011529432.1 35 5922
XM_011531131.1 XP_011529433.1 36 5919
GenBank mRNA AB191259.1 - 35 5824
AB191260.1 - 36 5853
AB371574.1 - 36 5853
AB371575.1 - 35 5824
AB371576.1 - 36 5853
AB371577.1 - 36 5848
AB371578.1 - 35 5824
AB371579.1 - 35 5824
AB593010.1 - 36 5988
AK074048.1 - 15 2301
AK090427.2 - 35 5890
AK300165.1 - 10 1161
AK304255.1 - 8 962
AL050396.1 - 9 1056
AL157419.1 - 13 1865
BC014654.1 - 7 767
BC067111.1 - 7 886
GU727643.1 - 37 6125
X53416.1 - 37 6048

View Full Product Details