Gene Transcripts

Gene Symbol
FLOT2
Entrez Gene ID
2319
Gene Name
flotillin 2
Gene Aliases
ECS-1, ECS1, ESA, ESA1, M17S1
Chromosome Location
Chr. 17: 28879339 - 28897728 on Build GRCh38
UniGene ID
Hs.514038
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004475.2 NP_004466.2 3 324
XM_017024394.1 XP_016879883.1 3 252
XM_017024395.1 XP_016879884.1 4 421
GenBank mRNA BC003683.1 - 3 236
BC017292.1 - 2 146
BT019477.1 - 1 54
BT019478.1 - 1 54
CA390100.1 - 3 324
CR541784.1 - 1 54
CR542265.1 - 1 54
M60922.1 - 3 180

View Full Product Details