Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001319053.1 | NP_001305982.1 | 9 | 1213 | |
NM_001470.3 | NP_001461.1 | 9 | 1277 | ||
NM_021903.2 | NP_068703.1 | 4 | 1077 | ||
NM_021904.3 | NP_068704.2 | 8 | 1091 | ||
XM_005248982.2 | XP_005249039.1 | 9 | 1092 | ||
XM_006715047.3 | XP_006715110.1 | 8 | 892 | ||
XM_011514453.2 | XP_011512755.1 | 7 | 871 | ||
XM_011514455.1 | XP_011512757.1 | 9 | 1092 | ||
XM_017010676.1 | XP_016866165.1 | 5 | 623 | ||
GenBank mRNA | AB208790.1 | - | 9 | 1012 | |
AF099148.1 | - | 9 | 1099 | ||
AF301005.1 | - | 8 | 994 | ||
AJ012185.1 | - | 8 | 991 | ||
AJ012186.1 | - | 4 | 640 | ||
AJ012187.1 | - | 7 | 805 | ||
AJ012288.1 | - | 9 | 1034 | ||
AJ225028.1 | - | 9 | 1225 | ||
AJ225029.1 | - | 4 | 819 | ||
AK223619.1 | - | 4 | 862 | ||
AK290191.1 | - | 4 | 862 | ||
AK312308.1 | - | 4 | 858 | ||
BC041332.2 | - | 4 | 862 | 5275467 | |
BC042598.1 | - | 4 | 851 | 4938491 | |
BC050532.1 | - | 4 | 849 | 5732186 | |
Y11044.1 | - | 8 | 988 |