Gene Transcripts
- Gene Symbol
- BLOC1S1
- Entrez Gene ID
- 2647
- Gene Name
- biogenesis of lysosomal organelles complex 1 subunit 1
- Gene Aliases
- BLOS1, BORCS1, GCN5L1, MICoA, RT14
- Chromosome Location
- Chr. 12: 55716034 - 55719707 on Build GRCh38
- UniGene ID
- Hs.600940
- Species
- Homo sapiens
- Gene Symbol
- BLOC1S1-RDH5
- Entrez Gene ID
- 100528022
- Gene Name
- BLOC1S1-RDH5 readthrough
- Gene Aliases
- Chromosome Location
- Chr. 12: 55716034 - 55724742 on Build GRCh38
- UniGene ID
- Hs.600940
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037658.1 |
|
1 |
138 |
|
GenBank mRNA |
AK096656.1 |
- |
1 |
120 |
|
AK307141.1 |
- |
1 |
122 |
|
View Full Product Details