Gene Transcripts

Gene Symbol
HEXA
Entrez Gene ID
3073
Gene Name
hexosaminidase subunit alpha
Gene Aliases
TSD
Chromosome Location
Chr. 15: 72343435 - 72376473 on Build GRCh38
UniGene ID
Hs.604479
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000520.5 NP_000511.2 9 1503
NM_001318825.1 NP_001305754.1 9 1536
NR_134869.1 9 1503
GenBank mRNA AK222502.1 - 9 1016
AK296528.1 - 9 874
AK297678.1 - 7 754
AK301000.1 - 9 1056
AK301138.1 - 7 875
AK307770.1 - 9 1023
AK310154.1 - 8 986
BC001138.2 - 7 643
BC018927.2 - 9 1013 3353424
BC021030.1 - 3 342
BC084537.1 - 9 1009 5459166
BX537989.1 - 8 6329
CR627386.1 - 9 1503
KU178109.1 - 9 1002
M13520.1 - 9 886

View Full Product Details