Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002191.3 | NP_002182.1 | 2 | 1225 | |
GenBank mRNA | AW025787.1 | - | 1 | 208 | |
BC006391.2 | - | 2 | 1162 | 4126990 | |
BT006954.1 | - | 2 | 1082 | ||
M13144.1 | - | 2 | 1039 | ||
M13981.1 | - | 2 | 1226 |