Gene Transcripts

Gene Symbol
KPNA1
Entrez Gene ID
3836
Gene Name
karyopherin subunit alpha 1
Gene Aliases
IPOA5, NPI-1, RCH2, SRP1
Chromosome Location
Chr. 3: 122421901 - 122514939 on Build GRCh38
UniGene ID
Hs.161008
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002264.3 NP_002255.3 2, 3 319
NR_026698.1 2, 3 319
XM_005247437.3 XP_005247494.1 2, 3 201
XM_011512797.2 XP_011511099.1 1, 2 136
XM_017006363.1 XP_016861852.1 2, 3 317
XM_017006364.1 XP_016861853.1 2, 3 196
GenBank mRNA AK299030.1 - 1, 2 205
AK312713.1 - 2, 3 314
BC002374.2 - 2, 3 308 2822859
BC003009.1 - 2, 3 308 2822859
BC090864.1 - 2, 3 290 6173605
BT006959.1 - 1, 2 131
CR456743.1 - 1, 2 131
DA308391.1 - 2, 3 319
DN991078.1 - 2, 3 161
S75295.1 - 2, 3 177

View Full Product Details