Gene Transcripts

Gene Symbol
MMP17
Entrez Gene ID
4326
Gene Name
matrix metallopeptidase 17
Gene Aliases
MMP-17, MT4-MMP, MT4MMP, MTMMP4
Chromosome Location
Chr. 12: 131828393 - 131851771 on Build GRCh38
UniGene ID
Hs.709245
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016155.5 NP_057239.4 9 1425
XM_011538355.2 XP_011536657.1 9 1279
XM_011538356.2 XP_011536658.1 9 1275
XM_011538357.2 XP_011536659.1 9 1224
GenBank mRNA AB021225.2 - 9 1431
BC040507.1 - 9 1417 5301089
BC045610.1 - 9 1434 5273795
BX389435.2 - 6 833
X89576.1 - 8 1303

View Full Product Details