Gene Transcripts

Gene Symbol
PC
Entrez Gene ID
5091
Gene Name
pyruvate carboxylase
Gene Aliases
PCB
Chromosome Location
Chr. 11: 66848522 - 66958418 on Build GRCh38
UniGene ID
Hs.89890
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000920.3 NP_000911.2 14 1887
NM_001040716.1 NP_001035806.1 15 2075
NM_022172.2 NP_071504.2 13 1842
XM_005274031.4 XP_005274088.1 13 1934
XM_005274032.4 XP_005274089.1 13 1816
XM_006718578.3 XP_006718641.1 13 1887
XM_006718579.3 XP_006718642.1 5 676
XM_011545086.2 XP_011543388.1 13 2203
XM_011545087.2 XP_011543389.1 5 801
XM_017017868.1 XP_016873357.1 16 2374
XM_017017869.1 XP_016873358.1 16 2245
XM_017017870.1 XP_016873359.1 15 2186
XM_017017871.1 XP_016873360.1 15 2057
XM_017017872.1 XP_016873361.1 14 2439
GenBank mRNA BC011617.2 - 14 1876
S72370.1 - 13 1875
U04641.1 - 13 1831
U30891.1 - 14 1845

View Full Product Details