Gene Transcripts

Gene Symbol
PEX1
Entrez Gene ID
5189
Gene Name
peroxisomal biogenesis factor 1
Gene Aliases
HMLR1, PBD1A, PBD1B, ZWS, ZWS1
Chromosome Location
Chr. 7: 92487023 - 92528531 on Build GRCh38
UniGene ID
Hs.164682
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000466.2 NP_000457.1 15 2617
NM_001282677.1 NP_001269606.1 14 2446
NM_001282678.1 NP_001269607.1 15 2652
XM_017012319.1 XP_016867808.1 10 1537
GenBank mRNA AB008112.1 - 15 2588
AB052090.1 - 14 2350
AB052091.1 - 15 2521
AB052093.1 - 15 2521
AB052094.1 - 15 2521
AB209355.1 - 4 2413
AF026086.1 - 15 2581
AF030356.1 - 15 2525
AK292955.1 - 15 2522
AK293757.1 - 15 2560
AK293848.1 - 9 1149
AK295686.1 - 12 1561
BC035575.1 - 15 2559 5498274
DB461678.1 - 2 291

View Full Product Details