Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001127218.2 | NP_001120690.1 | 8 | 1185 | |
NM_001256879.1 | NP_001243808.1 | 9 | 1546 | ||
NM_006230.3 | NP_006221.2 | 8 | 1012 | ||
XM_011515431.1 | XP_011513733.1 | 8 | 1008 | ||
GenBank mRNA | AK292347.1 | - | 9 | 1546 | |
AK298168.1 | - | 8 | 876 | ||
AK312294.1 | - | 8 | 1145 | ||
BC000459.2 | - | 8 | 975 | 2822169 | |
BC030001.2 | - | 7 | 952 | ||
U21090.1 | - | 8 | 974 |