Gene Transcripts
- Gene Symbol
- POU2F2
- Entrez Gene ID
- 5452
- Gene Name
- POU class 2 homeobox 2
- Gene Aliases
- OCT2, OTF2, Oct-2
- Chromosome Location
- Chr. 19: 42086110 - 42197931 on Build GRCh38
- UniGene ID
- Hs.147381
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001207025.2 |
NP_001193954.1
|
1, 2 |
92 |
|
NM_001207026.1 |
NP_001193955.1
|
1, 2 |
92 |
|
NM_001247994.1 |
NP_001234923.1
|
1, 2 |
92 |
|
NM_002698.4 |
NP_002689.1
|
1, 2 |
92 |
|
XM_005259010.3 |
XP_005259067.1
|
1, 2 |
109 |
|
XM_011527040.2 |
XP_011525342.1
|
1, 2 |
109 |
|
XM_011527041.2 |
XP_011525343.1
|
1, 2 |
109 |
|
XM_011527042.2 |
XP_011525344.1
|
1, 2 |
109 |
|
XM_011527043.2 |
XP_011525345.2
|
1, 2 |
109 |
|
XM_017026884.1 |
XP_016882373.1
|
1, 2 |
110 |
|
XM_017026891.1 |
XP_016882380.1
|
1, 2 |
110 |
|
XM_017026892.1 |
XP_016882381.1
|
1, 2 |
110 |
|
XM_017026893.1 |
XP_016882382.1
|
1, 2 |
109 |
|
XM_017026894.1 |
XP_016882383.1
|
1, 2 |
109 |
|
XM_017026896.1 |
XP_016882385.1
|
1, 2 |
110 |
|
GenBank mRNA |
BC006101.1 |
- |
1, 2 |
84 |
|
BT007438.1 |
- |
1, 2 |
30 |
|
CB995118.1 |
- |
1, 2 |
125 |
|
M36542.1 |
- |
1, 2 |
84 |
|
M36653.1 |
- |
1, 2 |
96 |
|
M36718.1 |
- |
1, 2 |
27 |
|
X13809.1 |
- |
1, 2 |
27 |
|
X13810.1 |
- |
1, 2 |
84 |
|
X53468.1 |
- |
1, 2 |
96 |
|
View Full Product Details