Gene Transcripts

Gene Symbol
MAPK13
Entrez Gene ID
5603
Gene Name
mitogen-activated protein kinase 13
Gene Aliases
MAPK 13, MAPK-13, PRKM13, SAPK4, p38delta
Chromosome Location
Chr. 6: 36130484 - 36144524 on Build GRCh38
UniGene ID
Hs.178695
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002754.4 NP_002745.1 1 129
NR_072996.1 1 129
GenBank mRNA AB209586.1 - 1 511
AB451221.1 - 1 30
AB451344.1 - 1 30
AF015256.1 - 1 90
AF092535.1 - 1 51
AF100546.1 - 1 32
AK222922.1 - 1 21
AK308018.1 - 1 110
BC000433.2 - 1 71 2819932
BC001641.2 - 1 71 2819932
BC004428.1 - 1 71 2819932
BC085196.1 - 1 99 6107307
BM921729.1 - 1 128
BT007221.1 - 1 30
CR536490.1 - 1 30
CR541687.1 - 1 30
DB473379.1 - 1 93
KU178281.1 - 1 30
KU178282.1 - 1 30
U93232.1 - 1 37
Y10488.1 - 1 52

View Full Product Details