Gene Transcripts

Gene Symbol
RBP1
Entrez Gene ID
5947
Gene Name
retinol binding protein 1
Gene Aliases
CRABP-I, CRBP, CRBP1, CRBPI, RBPC
Chromosome Location
Chr. 3: 139517434 - 139539829 on Build GRCh38
UniGene ID
Hs.529571
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130992.1 NP_001124464.1 2 406
NM_001130993.1 NP_001124465.1 2 406
NM_002899.3 NP_002890.2 2 406
GenBank mRNA AK290315.1 - 2 213
AK301684.1 - 2 296
AK307378.1 - 2 215
AK309492.1 - 2 406
BC121052.2 - 2 164 40118103
BU734286.1 - 3 507
CR541979.1 - 2 109
CR542005.1 - 2 109
DC356836.1 - 2 406
DC394326.1 - 2 296
M11433.1 - 2 234
M36809.1 - 2 181

View Full Product Details