Gene Transcripts

Gene Symbol
RFXAP
Entrez Gene ID
5994
Gene Name
regulatory factor X associated protein
Gene Aliases
Chromosome Location
Chr. 13: 36819202 - 36829603 on Build GRCh38
UniGene ID
Hs.24422
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000538.3 NP_000529.1 2 773
GenBank mRNA AK313912.1 - 2 755
BC026088.1 - 2 752 4827378

View Full Product Details