Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001128850.1 | NP_001122322.1 | 2, 3 | 533 | |
NM_004165.2 | NP_004156.1 | 2, 3 | 524 | ||
GenBank mRNA | BC011645.2 | - | 2, 3 | 515 | |
BC057815.1 | - | 2, 3 | 532 | 30341066 | |
L24564.1 | - | 2, 3 | 504 |