Gene Transcripts

Gene Symbol
SRSF1
Entrez Gene ID
6426
Gene Name
serine and arginine rich splicing factor 1
Gene Aliases
ASF, SF2, SF2p33, SFRS1, SRp30a
Chromosome Location
Chr. 17: 58000919 - 58007346 on Build GRCh38
UniGene ID
Hs.68714
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001078166.1 NP_001071634.1 1 280
NM_006924.4 NP_008855.1 1 280
NR_034041.1 1 280
GenBank mRNA AB062124.1 - 1 161
AB209558.1 - 1 167
AK225711.1 - 1 158
AK289933.1 - 1 189
AK312781.1 - 1 177
BC010264.1 - 1 106 2900101
BC033785.1 - 1 166 5166554
DA939911.1 - 1 280
M69040.1 - 1 195
M72709.1 - 1 167

View Full Product Details