Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000340.1 | NP_000331.1 | 3 | 584 | |
NM_001278659.1 | NP_001265588.1 | 3 | 584 | ||
XM_011513087.2 | XP_011511389.1 | 4 | 331 | ||
GenBank mRNA | AK290846.1 | - | 3 | 327 | |
AK292741.1 | - | 3 | 327 | ||
AK313622.1 | - | 3 | 325 | ||
BC060041.1 | - | 3 | 329 | 4716156 | |
J03810.1 | - | 3 | 313 |