Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005994.3 | NP_005985.3 | 6, 7 | 1977 | |
XM_011525159.1 | XP_011523461.1 | 5, 6 | 1648 | ||
GenBank mRNA | AB209378.1 | - | 5 | 4155 | |
AK001031.1 | - | 1 | 814 | ||
AK127429.1 | - | 2 | 2765 | ||
AL832900.1 | - | 6, 7 | 1517 | ||
BC052566.1 | - | 6, 7 | 1977 | 6339405 | |
BC070054.1 | - | 6, 7 | 1942 | ||
U28049.1 | - | 6, 7 | 1713 |