Gene Transcripts
- Gene Symbol
- PPFIA4
- Entrez Gene ID
- 8497
- Gene Name
- PTPRF interacting protein alpha 4
- Gene Aliases
- Chromosome Location
- Chr. 1: 203026491 - 203078736 on Build GRCh38
- UniGene ID
- Hs.153648
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001304331.1 |
NP_001291260.1
|
19, 20 |
2914 |
|
NM_001304332.1 |
NP_001291261.1
|
19, 20 |
2914 |
|
XM_006711586.2 |
XP_006711649.1
|
19, 20 |
2944 |
|
XM_011510070.2 |
XP_011508372.1
|
7, 8 |
973 |
|
XM_017002606.1 |
XP_016858095.1
|
19, 20 |
2944 |
|
XM_017002607.1 |
XP_016858096.1
|
18, 19 |
2809 |
|
XM_017002608.1 |
XP_016858097.1
|
19, 20 |
2944 |
|
XM_017002609.1 |
XP_016858098.1
|
18, 19 |
2809 |
|
GenBank mRNA |
AB020704.1 |
- |
1, 2 |
43 |
|
AF034801.1 |
- |
4, 5 |
465 |
|
AK023365.1 |
- |
1, 2 |
111 |
|
AK126624.1 |
- |
6, 7 |
1482 |
|
AK295757.1 |
- |
13, 14 |
1642 |
|
BC132923.1 |
- |
6, 7 |
1424 |
40146945 |
BC132925.1 |
- |
6, 7 |
1424 |
40146947 |
BC144262.1 |
- |
6, 7 |
1424 |
9052782 |
EF428334.1 |
- |
24, 25 |
2781 |
|
View Full Product Details