Gene Transcripts

Gene Symbol
CTDP1
Entrez Gene ID
9150
Gene Name
CTD phosphatase subunit 1
Gene Aliases
CCFDN, FCP1
Chromosome Location
Chr. 18: 79679801 - 79754510 on Build GRCh38
UniGene ID
Hs.465490
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001202504.1 NP_001189433.1 7 718
NM_001318511.1 NP_001305440.1 7 1091
NM_004715.4 NP_004706.3 7 1091
NM_048368.3 NP_430255.2 7 1091
XM_011526261.1 XP_011524563.1 7 1069
XM_017026078.1 XP_016881567.1 5 571
GenBank mRNA AF081287.1 - 6 639
AF154115.1 - 7 1027
BC015010.1 - 1 38
BC032515.1 - 7 745
BC047999.1 - 7 1091
BC052576.1 - 7 683
BC063447.1 - 7 1046 5500032

View Full Product Details