Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005454.2 | NP_005445.1 | 1, 2 | 544 | |
GenBank mRNA | AF400435.1 | - | 1, 2 | 499 | |
BC069371.1 | - | 1, 2 | 499 | ||
BC069405.1 | - | 1, 2 | 499 | 7262136 | |
BC069491.1 | - | 1, 2 | 499 | 7262148 | |
BC069503.1 | - | 1, 2 | 499 | 7262160 | |
BC103976.1 | - | 1, 2 | 499 | 40015380 | |
BC103977.1 | - | 1, 2 | 499 | 40015381 | |
CD652441.1 | - | 1, 2 | 564 |