Gene Transcripts
- Gene Symbol
- MYOT
- Entrez Gene ID
- 9499
- Gene Name
- myotilin
- Gene Aliases
- LGMD1, LGMD1A, MFM3, TTID, TTOD
- Chromosome Location
- Chr. 5: 137867282 - 137887851 on Build GRCh38
- UniGene ID
- Hs.84665
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001300911.1 |
NP_001287840.1
|
3, 4 |
304 |
|
NM_006790.2 |
NP_006781.1
|
2, 3 |
655 |
|
XM_017010060.1 |
XP_016865549.1
|
3, 4 |
878 |
|
XM_017010061.1 |
XP_016865550.1
|
1, 2 |
311 |
|
GenBank mRNA |
AF133820.2 |
- |
2, 3 |
640 |
|
AF144477.1 |
- |
2, 3 |
626 |
|
AK300076.1 |
- |
3, 4 |
284 |
|
AK307620.1 |
- |
3, 4 |
518 |
|
AK312733.1 |
- |
2, 3 |
635 |
|
BC005376.1 |
- |
2, 3 |
635 |
3934230 |
KU178476.1 |
- |
1, 2 |
346 |
|
View Full Product Details