Gene Transcripts

Gene Symbol
SLC23A2
Entrez Gene ID
9962
Gene Name
solute carrier family 23 member 2
Gene Aliases
NBTL1, SLC23A1, SVCT2, YSPL2, hSVCT2
Chromosome Location
Chr. 20: 4852356 - 5010293 on Build GRCh38
UniGene ID
Hs.516866
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005116.5 NP_005107.4 15 1926
NM_203327.1 NP_976072.1 15 1944
XM_011529414.2 XP_011527716.1 15 1931
XM_011529415.2 XP_011527717.1 14 1752
XM_011529416.2 XP_011527718.1 14 2061
XM_011529417.2 XP_011527719.1 14 1808
XM_017028171.1 XP_016883660.1 15 1965
XM_017028172.1 XP_016883661.1 15 1969
XM_017028173.1 XP_016883662.1 15 1949
XM_017028174.1 XP_016883663.1 15 1844
XM_017028175.1 XP_016883664.1 11 1280
XM_017028176.1 XP_016883665.1 12 1598
XM_017028177.1 XP_016883666.1 12 1585
XM_017028178.1 XP_016883667.1 14 1851
XM_017028179.1 XP_016883668.1 14 1838
XM_017028180.1 XP_016883669.1 10 1162
GenBank mRNA AF058319.1 - 13 1551
AF092511.1 - 14 1847
AF164142.1 - 15 1944
AJ269478.1 - 13 1551
AJ292318.1 - 14 1782
AK296304.1 - 12 1592
AY380556.1 - 13 1551
D87075.2 - 15 1926
EF032501.1 - 13 1551

View Full Product Details