Gene Transcripts

Gene Symbol
DOPEY2
Entrez Gene ID
9980
Gene Name
dopey family member 2
Gene Aliases
21orf5, C21orf5
Chromosome Location
Chr. 21: 36156781 - 36294274 on Build GRCh38
UniGene ID
Hs.204575
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320714.1 NP_001307643.1 27, 28 5823
NM_005128.3 NP_005119.2 27, 28 5714
XM_017028509.1 XP_016883998.1 24, 25 5217
GenBank mRNA AB023150.2 - 24, 25 5183
AJ001857.1 - 1, 2 154
AJ237839.2 - 27, 28 5720
AK026478.1 - 6, 7 650

View Full Product Details