Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001297764.1 | NP_001284693.1 | 4 | 451 | |
NM_005709.3 | NP_005700.2 | 4 | 451 | ||
NM_153676.3 | NP_710142.1 | 4 | 451 | ||
NR_123738.1 | 4 | 451 | |||
XM_011519832.2 | XP_011518134.1 | 4 | 451 | ||
XM_011519834.2 | XP_011518136.1 | 4 | 451 | ||
XM_017017072.1 | XP_016872561.1 | 4 | 451 | ||
XM_017017073.1 | XP_016872562.1 | 4 | 451 | ||
XM_017017074.1 | XP_016872563.1 | 4 | 451 | ||
XM_017017075.1 | XP_016872564.1 | 4 | 451 | ||
GenBank mRNA | AB006955.1 | - | 3 | 494 | |
AB018687.1 | - | 4 | 431 | ||
AF039699.1 | - | 4 | 438 | ||
AF039700.1 | - | 4 | 438 | ||
AK225614.1 | - | 4 | 451 | ||
AK290788.1 | - | 4 | 403 | ||
BC016057.1 | - | 4 | 435 | 4867369 | |
KU178498.1 | - | 4 | 342 | ||
KU178499.1 | - | 4 | 342 |