Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005877.5 | NP_005868.1 | 9 | 1443 | |
GenBank mRNA | AK098463.1 | - | 2 | 209 | |
AK303273.1 | - | 10 | 1368 | ||
BC001976.1 | - | 9 | 1383 | 3459964 | |
BC007684.2 | - | 9 | 1410 | 3634035 | |
CR456575.1 | - | 9 | 1386 | ||
X85237.1 | - | 9 | 1385 |