Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001323041.1 | NP_001309970.1 | 5 | 966 | |
NM_001323042.1 | NP_001309971.1 | 5 | 450 | ||
NM_001323043.1 | NP_001309972.1 | 5 | 851 | ||
NM_001323044.1 | NP_001309973.1 | 5 | 1224 | ||
NM_001323045.1 | NP_001309974.1 | 6 | 935 | ||
NM_001323047.1 | NP_001309976.1 | 5 | 966 | ||
NM_001323048.1 | NP_001309977.1 | 5 | 851 | ||
NM_001323049.1 | NP_001309978.1 | 4 | 1140 | ||
NM_001323050.1 | NP_001309979.1 | 6 | 935 | ||
NM_001323051.1 | NP_001309980.1 | 5 | 1224 | ||
NM_001323052.1 | NP_001309981.1 | 5 | 851 | ||
NM_001323053.1 | NP_001309982.1 | 5 | 966 | ||
NM_006608.2 | NP_006599.2 | 4 | 755 | ||
NR_136564.1 | 6 | 1050 | |||
NR_136565.1 | 5 | 851 | |||
NR_136566.1 | 5 | 966 | |||
NR_136567.1 | 5 | 851 | |||
NR_136568.1 | 6 | 935 | |||
GenBank mRNA | AJ011863.1 | - | 4 | 334 | |
AK294749.1 | - | 4 | 481 | ||
BC002447.2 | - | 5 | 899 | 3346990 | |
CX763183.1 | - | 4 | 631 |