Gene Transcripts

Gene Symbol
PNRC1
Entrez Gene ID
10957
Gene Name
proline rich nuclear receptor coactivator 1
Gene Aliases
B4-2, PNAS-145, PROL2, PRR2
Chromosome Location
Chr. 6: 89080710 - 89085160 on Build GRCh38
UniGene ID
Hs.75969
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006813.2 NP_006804.1 2 779
GenBank mRNA AK130046.1 - 2 737
AK130722.1 - 1 819
AK312665.1 - 2 738
AY303779.1 - 2 357
BC018112.1 - 2 649 3916954
BC044919.1 - 2 959
U03105.1 - 2 707

View Full Product Details