Gene Transcripts

Gene Symbol
LMAN2
Entrez Gene ID
10960
Gene Name
lectin, mannose binding 2
Gene Aliases
C5orf8, GP36B, VIP36
Chromosome Location
Chr. 5: 177331562 - 177351884 on Build GRCh38
UniGene ID
Hs.75864
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006816.2 NP_006807.1 6, 7 1029
GenBank mRNA AK292099.1 - 6, 7 806
AK301607.1 - 7, 8 713
AK308027.1 - 6, 7 940
BC017263.2 - 6, 7 809 4135210
U10362.1 - 6, 7 792

View Full Product Details