Gene Transcripts

Gene Symbol
WDR45
Entrez Gene ID
11152
Gene Name
WD repeat domain 45
Gene Aliases
JM5, NBIA4, NBIA5, WDRX1, WIPI-4, WIPI4
Chromosome Location
Chr. X: 49074433 - 49101121 on Build GRCh38
UniGene ID
Hs.632807
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029896.1 NP_001025067.1 7 640
NM_007075.3 NP_009006.2 8 910
GenBank mRNA AJ005897.1 - 7 596
AK225122.1 - 7 619
AK315717.1 - 7 627
AY691428.1 - 6 468
BC000464.2 - 7 634 2822742
BC003037.1 - 7 634 2822742
BC009027.1 - 4 297
BC069206.1 - 8 910 5801768
BQ068083.1 - 7 640
CR456725.1 - 6 468

View Full Product Details