Gene Transcripts

Gene Symbol
NISCH
Entrez Gene ID
11188
Gene Name
nischarin
Gene Aliases
I-1, IR1, IRAS, hIRAS
Chromosome Location
Chr. 3: 52455508 - 52493072 on Build GRCh38
UniGene ID
Hs.435290
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276293.1 NP_001263222.1 1, 2 216
NM_001276294.1 NP_001263223.1 1, 2 216
NM_007184.3 NP_009115.2 1, 2 216
GenBank mRNA AB023192.1 - 2, 3 154
AF082516.1 - 1, 2 108
AK027428.1 - 1, 2 94
AK291398.1 - 1, 2 120
AK297657.1 - 1, 2 120
AK315001.1 - 1, 2 120
AL703940.1 - 1, 2 90
BC038102.2 - 1, 2 120 5268247
BC054494.1 - 1, 2 114 6195532
BC056900.1 - 1, 2 86 6143316
DB496030.1 - 1, 2 216

View Full Product Details