Gene Transcripts
- Gene Symbol
- ARHGEF15
- Entrez Gene ID
- 22899
- Gene Name
- Rho guanine nucleotide exchange factor 15
- Gene Aliases
- ARGEF15, E5, Ephexin5, Vsm-RhoGEF
- Chromosome Location
- Chr. 17: 8310238 - 8322516 on Build GRCh38
- UniGene ID
- Hs.443109
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025014.1 |
NP_079290.1
|
14, 15 |
2352 |
|
NM_173728.3 |
NP_776089.2
|
14, 15 |
2416 |
|
XM_011523734.1 |
XP_011522036.1
|
14, 15 |
2461 |
|
XM_011523735.1 |
XP_011522037.1
|
14, 15 |
2397 |
|
XM_011523736.1 |
XP_011522038.1
|
14, 15 |
2412 |
|
XM_011523737.1 |
XP_011522039.1
|
11, 12 |
1453 |
|
GenBank mRNA |
AB020722.1 |
- |
14, 15 |
2332 |
|
AK291626.1 |
- |
14, 15 |
2376 |
|
AK300327.1 |
- |
14, 15 |
1837 |
|
BC036749.1 |
- |
14, 15 |
2371 |
5204409 |
BQ877184.1 |
- |
1, 2 |
90 |
|
KU178557.1 |
- |
13, 14 |
2261 |
|
KU178558.1 |
- |
12, 13 |
2108 |
|
View Full Product Details