Gene Transcripts

Gene Symbol
CARD8
Entrez Gene ID
22900
Gene Name
caspase recruitment domain family member 8
Gene Aliases
CARDINAL, DACAR, DAKAR, NDPP, NDPP1, TUCAN
Chromosome Location
Chr. 19: 48203148 - 48256269 on Build GRCh38
UniGene ID
Hs.446146
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184900.1 NP_001171829.1 9 1290
NM_001184901.1 NP_001171830.1 9 1237
NM_014959.3 NP_055774.2 10 1348
NR_033678.1 8 1207
NR_033679.1 8 1249
XM_006723091.3 XP_006723154.1 11 1501
XM_006723092.3 XP_006723155.1 13 2039
XM_006723093.3 XP_006723156.1 11 1822
XM_006723094.3 XP_006723157.1 11 1481
XM_006723095.3 XP_006723158.1 11 1468
XM_006723096.3 XP_006723159.1 12 1890
XM_006723097.3 XP_006723160.1 10 1673
XM_006723098.3 XP_006723161.1 10 1460
XM_006723099.3 XP_006723162.1 9 1346
XM_006723101.2 XP_006723164.1 10 1363
XM_006723102.3 XP_006723165.1 9 1303
XM_006723104.3 XP_006723167.1 10 1360
XM_011526641.2 XP_011524943.1 14 2346
XM_011526642.2 XP_011524944.1 13 2241
XM_011526643.2 XP_011524945.1 12 1928
XM_011526644.2 XP_011524946.1 10 1513
XM_011526646.2 XP_011524948.1 11 1987
XM_011526647.2 XP_011524949.1 10 1876
XM_011526648.2 XP_011524950.1 11 1993
XM_017026482.1 XP_016881971.1 11 1826
XM_017026483.1 XP_016881972.1 11 1779
XM_017026484.1 XP_016881973.1 11 1787
XM_017026485.1 XP_016881974.1 11 1484
XM_017026486.1 XP_016881975.1 9 1343
XM_017026487.1 XP_016881976.1 8 1712
XM_017026488.1 XP_016881977.1 12 1928
XM_017026489.1 XP_016881978.1 11 1513
GenBank mRNA AB023172.1 - 8 1242
AF143869.1 - 1 29
AF322184.1 - 8 1242
AF331519.1 - 6 929
AF405558.1 - 6 929
AK297069.1 - 9 1227
AK302327.1 - 8 1177
AY026322.1 - 6 929
EU118120.1 - 9 1290
EU118121.1 - 8 1249
EU118122.1 - 6 1090

View Full Product Details