Gene Transcripts

Gene Symbol
COLGALT2
Entrez Gene ID
23127
Gene Name
collagen beta(1-O)galactosyltransferase 2
Gene Aliases
C1orf17, GLT25D2
Chromosome Location
Chr. 1: 183929662 - 184051612 on Build GRCh38
UniGene ID
Hs.387995
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303420.1 NP_001290349.1 10 1703
NM_001303421.1 NP_001290350.1 10 1217
NM_015101.3 NP_055916.1 10 1703
XM_011509332.2 XP_011507634.1 11 1509
XM_011509334.2 XP_011507636.1 11 1316
XM_011509335.2 XP_011507637.1 11 1367
XM_011509336.2 XP_011507638.1 11 1144
XM_011509337.2 XP_011507639.1 7 1443
XM_011509338.2 XP_011507640.1 8 1195
XM_017000750.1 XP_016856239.1 10 1268
XM_017000751.1 XP_016856240.1 5 1127
GenBank mRNA AB011156.1 - 10 1628
AF288389.1 - 10 1662
AK095090.1 - 5 938
AK095192.1 - 10 1191
AK127287.1 - 2 1208
AK293973.1 - 10 1410
BC035672.1 - 10 1685 5740842

View Full Product Details