Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012213.2 | NP_036345.2 | 5 | 1071 | |
GenBank mRNA | AF090834.1 | - | 5 | 1051 | |
AF097832.2 | - | 5 | 989 | ||
AF153679.1 | - | 5 | 1029 | ||
AK094095.1 | - | 1 | 1994 | ||
BC000286.1 | - | 5 | 1023 | 3357140 | |
BC052592.1 | - | 5 | 1071 | 6421051 |