Gene Transcripts

Gene Symbol
TARDBP
Entrez Gene ID
23435
Gene Name
TAR DNA binding protein
Gene Aliases
ALS10, TDP-43
Chromosome Location
Chr. 1: 11012622 - 11030528 on Build GRCh38
UniGene ID
Hs.300624
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007375.3 NP_031401.1 3 405
XM_017000863.1 XP_016856352.1 3 393
XM_017000864.1 XP_016856353.1 3 393
XM_017000865.1 XP_016856354.1 3 393
XM_017000866.1 XP_016856355.1 3 393
XM_017000867.1 XP_016856356.1 3 393
XM_017000868.1 XP_016856357.1 3 393
GenBank mRNA AK222754.1 - 3 343
AK299454.1 - 3 156
AK312416.1 - 3 373
AL050265.1 - 3 359
BC001487.2 - 3 358
BC071657.1 - 3 356 5498250
BC095435.1 - 3 357 30389805
BI464234.1 - 4 319
CD703666.1 - 3 431
CR533534.1 - 2 271
EF434181.1 - 2 271
EF434182.1 - 2 271
HQ628634.1 - 3 180
HQ628635.1 - 2 271
HQ628636.1 - 2 271
HQ628637.1 - 2 271
U23731.1 - 2 271

View Full Product Details