Gene Transcripts

Gene Symbol
MTO1
Entrez Gene ID
25821
Gene Name
mitochondrial tRNA translation optimization 1
Gene Aliases
CGI-02, COXPD10
Chromosome Location
Chr. 6: 73461727 - 73501456 on Build GRCh38
UniGene ID
Hs.347614
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001123226.1 NP_001116698.1 11 1907
NM_012123.3 NP_036255.2 10 1787
NM_133645.2 NP_598400.1 11 1862
XM_006715444.2 XP_006715507.1 10 1684
XM_006715445.2 XP_006715508.1 10 1489
XM_006715446.3 XP_006715509.1 10 1787
XM_011535723.1 XP_011534025.1 11 1876
XM_011535724.1 XP_011534026.1 11 1744
XM_011535725.2 XP_011534027.1 11 1612
XM_017010697.1 XP_016866186.1 10 1684
XM_017010698.1 XP_016866187.1 11 1876
XM_017010699.1 XP_016866188.1 9 1209
XM_017010700.1 XP_016866189.1 9 1481
GenBank mRNA AF132937.1 - 10 1709
AF319422.2 - 10 1753
AF469110.1 - 9 1462
AF469111.1 - 9 1843
AJ420530.1 - 9 1696
AK026073.1 - 6 771
AK074625.1 - 10 1766
AK225828.1 - 11 1891
AL833823.1 - 10 1631
AY078985.1 - 11 1828
AY078986.1 - 11 1828
BC005808.2 - 11 1547
BC011051.1 - 10 1728 4339769

View Full Product Details