Gene Transcripts

Gene Symbol
HECTD1
Entrez Gene ID
25831
Gene Name
HECT domain E3 ubiquitin protein ligase 1
Gene Aliases
EULIR
Chromosome Location
Chr. 14: 31100115 - 31208060 on Build GRCh38
UniGene ID
Hs.708017
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015382.3 NP_056197.3 20 3589
XM_005267502.2 XP_005267559.2 20 3751
XM_011536619.2 XP_011534921.1 20 3187
XM_011536621.2 XP_011534923.1 20 3187
XM_011536622.1 XP_011534924.1 11 1514
XM_017021148.1 XP_016876637.1 19 3081
XM_017021149.1 XP_016876638.1 19 3081
XM_017021150.1 XP_016876639.1 19 3081
XM_017021151.1 XP_016876640.1 20 3783
XM_017021152.1 XP_016876641.1 20 3783
XM_017021153.1 XP_016876642.1 20 3187
GenBank mRNA AB032957.2 - 12 1735
AK095634.1 - 11 1482
AY254380.1 - 20 3389

View Full Product Details