Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001244682.1 | NP_001231611.1 | 10 | 1081 | |
NM_014352.3 | NP_055167.2 | 10 | 1191 | ||
XM_011542739.2 | XP_011541041.1 | 12 | 1406 | ||
XM_011542740.2 | XP_011541042.1 | 11 | 1264 | ||
XM_011542741.2 | XP_011541043.1 | 9 | 1115 | ||
XM_011542742.2 | XP_011541044.1 | 11 | 1369 | ||
XM_011542743.2 | XP_011541045.1 | 9 | 3510 | ||
XM_017017487.1 | XP_016872976.1 | 10 | 1191 | ||
GenBank mRNA | AF133895.1 | - | 10 | 1048 | |
AJ012214.1 | - | 9 | 1041 | ||
AK097382.1 | - | 3 | 570 | ||
AK291993.1 | - | 10 | 1191 | ||
AK302212.1 | - | 10 | 1081 | ||
BC101649.1 | - | 10 | 1126 | 8069155 | |
BC143593.1 | - | 11 | 1173 | ||
BC143594.1 | - | 11 | 1170 | ||
BC143595.1 | - | 11 | 1171 | ||
CU451853.1 | - | 2 | 177 |