Gene Transcripts

Gene Symbol
METTL21B
Entrez Gene ID
25895
Gene Name
methyltransferase like 21B
Gene Aliases
FAM119B
Chromosome Location
Chr. 12: 57772600 - 57782541 on Build GRCh38
UniGene ID
Hs.632720
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015433.2 NP_056248.2 1 215
NM_206914.1 NP_996797.1 1 215
GenBank mRNA AF455816.1 - 1 215
AK024983.1 - 1 159
AK292276.1 - 1 159
AL050100.1 - 1 137
BC016395.1 - 1 215 3882414
BC099841.1 - 1 190 40010149
BC103501.1 - 1 190 40010152

View Full Product Details