Gene Transcripts

Gene Symbol
LSM1
Entrez Gene ID
27257
Gene Name
LSM1 homolog, mRNA degradation associated
Gene Aliases
CASM, YJL124C
Chromosome Location
Chr. 8: 38163321 - 38176730 on Build GRCh38
UniGene ID
Hs.425311
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014462.2 NP_055277.1 4 781
NR_045492.1 3 665
NR_045493.1 4 503
GenBank mRNA AF000177.1 - 4 535
AJ238094.1 - 4 559
AK312159.1 - 4 534
BC001767.1 - 4 468 3353833
BE670269.1 - 1 383
BU521199.1 - 3 430
KU178606.1 - 4 371
KU178607.1 - 3 255

View Full Product Details