Gene Transcripts

Gene Symbol
C11orf54
Entrez Gene ID
28970
Gene Name
chromosome 11 open reading frame 54
Gene Aliases
PTD012, PTOD012
Chromosome Location
Chr. 11: 93741594 - 93765323 on Build GRCh38
UniGene ID
Hs.8360
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286067.1 NP_001272996.1 7 830
NM_001286068.1 NP_001272997.1 6 693
NM_001286069.1 NP_001272998.1 6 698
NM_001286070.1 NP_001272999.1 5 546
NM_014039.3 NP_054758.2 6 698
XM_006718824.1 XP_006718887.1 7 739
XM_011542781.2 XP_011541083.1 6 1012
XM_011542782.2 XP_011541084.1 6 1103
XM_017017613.1 XP_016873102.1 7 4154
XM_017017614.1 XP_016873103.1 7 981
XM_017017615.1 XP_016873104.1 7 1434
XM_017017616.1 XP_016873105.1 6 690
XM_017017619.1 XP_016873108.1 5 544
GenBank mRNA AF092133.1 - 6 606
AK129932.1 - 1 117
AK225977.1 - 6 628
AK291833.1 - 6 604
AK292215.1 - 7 749
AL136605.1 - 6 624
AY203960.1 - 5 574
BC007110.1 - 5 453
BC012298.1 - 6 665 3852558
BI597310.1 - 5 468
CR533538.1 - 5 466

View Full Product Details