Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001277198.1 | NP_001264127.1 | 4 | 427 | |
NM_001277199.1 | NP_001264128.1 | 4 | 752 | ||
NM_001277200.1 | NP_001264129.1 | 4 | 474 | ||
NM_014184.3 | NP_054903.1 | 4 | 474 | ||
NR_102347.1 | 3 | 455 | |||
GenBank mRNA | AF161512.1 | - | 4 | 393 | |
AK024569.1 | - | 389 | |||
AK222634.1 | - | 4 | 379 | ||
AK312064.1 | - | 4 | 399 | ||
BC000573.1 | - | 4 | 389 | 3163724 | |
BI752106.1 | - | 4 | 343 | ||
BQ051754.1 | - | 4 | 663 | ||
BQ575976.1 | - | 2 | 428 | ||
DA185135.1 | - | 4 | 475 | ||
HY018269.1 | - | 3 | 371 | ||
HY145928.1 | - | 4 | 399 |