Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020369.2 | NP_065102.1 | 2 | 849 | |
GenBank mRNA | AF281049.1 | - | 2 | 849 | |
AK225867.1 | - | 2 | 803 | ||
AK300518.1 | - | 3 | 893 | ||
AK314050.1 | - | 2 | 754 | ||
BC035606.1 | - | 2 | 724 | 5172588 |