Gene Transcripts

Gene Symbol
NDUFAF1
Entrez Gene ID
51103
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 1
Gene Aliases
CGI-65, CGI65, CIA30
Chromosome Location
Chr. 15: 41387349 - 41402694 on Build GRCh38
UniGene ID
Hs.106529
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016013.3 NP_057097.2 2 420
NR_045620.1 2 420
XM_006720555.2 XP_006720618.1 2 350
XM_011521658.1 XP_011519960.1 2 284
XM_011521659.2 XP_011519961.1 2 447
XM_017022293.1 XP_016877782.1 2 351
GenBank mRNA AK309889.1 - 2 420
BC000780.2 - 2 404 3504355

View Full Product Details