Gene Transcripts

Gene Symbol
PHF20
Entrez Gene ID
51230
Gene Name
PHD finger protein 20
Gene Aliases
C20orf104, GLEA2, HCA58, NZF, TDRD20A, TZP
Chromosome Location
Chr. 20: 35771999 - 35950377 on Build GRCh38
UniGene ID
Hs.517044
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016436.4 NP_057520.2 4 395
XM_011528843.1 XP_011527145.1 4 384
XM_017027864.1 XP_016883353.1 5 477
XM_017027865.1 XP_016883354.1 5 678
XM_017027866.1 XP_016883355.1 4 598
XM_017027867.1 XP_016883356.1 4 433
XM_017027869.1 XP_016883358.1 5 472
XM_017027870.1 XP_016883359.1 5 478
XM_017027871.1 XP_016883360.1 5 478
GenBank mRNA AF220416.1 - 4 359
AF258787.1 - 4 381
AF348207.1 - 3 320
AK054597.1 - 5 652
AK314503.1 - 4 359
AL558423.3 - 3 318
AY027523.1 - 3 320
AY134747.1 - 4 372
BC006415.1 - 4 335
BC016948.1 - 4 348
BC048210.1 - 5 428
BC080598.1 - 4 359
BC093405.1 - 4 395
BC150178.1 - 4 361 40148074
BG778940.1 - 5 465
BM906582.1 - 4 384
BX643875.1 - 4 353
CD690482.1 - 4 397
L09749.1 - 3 276

View Full Product Details