Gene Transcripts

Gene Symbol
KRCC1
Entrez Gene ID
51315
Gene Name
lysine rich coiled-coil 1
Gene Aliases
CHBP2
Chromosome Location
Chr. 2: 88027203 - 88055801 on Build GRCh38
UniGene ID
Hs.469254
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304526.1 NP_001291455.1 4 619
NM_016618.2 NP_057702.1 4 680
XM_017004292.1 XP_016859781.1 4 981
GenBank mRNA AF208845.1 - 4 608
AF217520.1 - 4 619
AK025986.1 - 358
BC015927.1 - 4 571 3908532
BC107580.1 - 4 646 4668988
EF413023.1 - 1 214

View Full Product Details